  • Search Results
  • Improving the distribution of Doxil® in the tumor matrix by depletion of tumor hyaluronan.
Improving the distribution of Doxil® in the tumor matrix by depletion of tumor hyaluronan.
Journal of controlled release : official journal of the Controlled Release Society (2014-05-24)
Aditya G Kohli, Saul Kivimäe, Matthew R Tiffany, Francis C Szoka
ABSTRACT
Liposomes improve the pharmacokinetics and safety of rapidly cleared drugs, but have not yet improved the clinical efficacy compared to the non-encapsulated drug. This inability to improve efficacy may be partially due to the non-uniform distribution of liposomes in solid tumors. The tumor extra-cellular matrix is a barrier to distribution and includes the high molecular weight glycosaminoglycan, hyaluronan (HA). Strategies to remove HA or block its synthesis may improve drug delivery into solid tumors. Orally administered methylumbelliferone (MU) is an inhibitor of HA synthesis, but it is limited by low potency and limited solubility. In this study, we encapsulate a water-soluble phosphorylated prodrug of MU (MU-P) in a liposome (L-MU-P). We demonstrate that L-MU-P is a more potent inhibitor of HA synthesis than oral MU in the 4T1 murine mammary carcinoma model using both a quantitative ELISA and histochemistry. We show that HA depletion improves the tumor distribution of liposomes computed using Mander's colocalization analysis of liposomes with the tumor vasculature. Hyaluronan depletion also increases the fraction of the tumor area positive for liposomes. This improved distribution extends the overall survival of mice treated with Doxil®.
Sigma-Aldrich
O-Phosphorylethanolamine
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, Vetec, reagent grade, ≥92.5%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Umbelliferone, analytical standard
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, Vetec, reagent grade, 99%
Sigma-Aldrich
L-Lysine monohydrochloride, Vetec, reagent grade, ≥98%