  • Search Results
  • Green synthesis and synergistic catalytic effect ofAg/reduced graphene oxide nanocomposite.
Green synthesis and synergistic catalytic effect ofAg/reduced graphene oxide nanocomposite.
Nanoscale research letters (2014-09-27)
Kai-Chih Hsu, Dong-Hwang Chen
ABSTRACT
A nanocomposite of silver nanoparticles and reduced graphene oxide (Ag/rGO) has been developed as a catalyst for the reduction of 4-nitrophenol (4-NP) to 4-aminophenol (4-AP) with sodium borohydride, owing to the larger specific surface area and synergistic effect of rGO. A facile and rapid microwave-assisted green route has been used for the uniform deposition of Ag nanoparticles and the reduction of graphene oxide simultaneously with l-arginine as the reducing agent. The resulting Ag/rGO nanocomposite contained about 51 wt% of Ag, and the Ag nanoparticles deposited on the surface of rGO had a mean diameter of 8.6 ± 3.5 nm. Also, the Ag/rGO nanocomposite exhibited excellent catalytic activity and stability toward the reduction of 4-NP to 4-AP with sodium borohydride. The reduction reaction obeyed the pseudo-first-order kinetics. The rate constants increased not only with the increase of temperature and catalyst amount but also with the increase of initial 4-NP concentration, revealing that the support rGO could enhance the catalytic activity via a synergistic effect. A mechanism for the catalytic reduction of 4-NP with NaBH4 by Ag/rGO nanocomposite via both the liquid-phase and solid-phase routes has been suggested.
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Sodium nitrate, analytical standard
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Supelco
4-Nitrophenol
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Supelco
4-Nitrophenol solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Sodium nitrate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%