  • Search Results
  • Application the mass spectrometry MALDI-TOF technique for detection of Babesia canis canis infection in dogs.
Application the mass spectrometry MALDI-TOF technique for detection of Babesia canis canis infection in dogs.
Parasitology research (2014-09-23)
Łukasz Adaszek, Tomasz Banach, Michał Bartnicki, Dagmara Winiarczyk, Paweł Łyp, Stanisław Winiarczyk
ABSTRACT
The aim of this study was to use rapid mass spectrometry (MS)-based proteomics analyses for diagnosis of Babesia canis canis infections in dogs. The study was conducted on two groups of dogs--healthy dogs and dogs infected with B. canis canis which demonstrated symptoms of babesiosis. The matrix-assisted laser desorption ionization time-of-flight (MALDI-TOF) MS technique revealed the presence of a protein fraction of 51-52 kDa in the blood serum of all the animals infected with the protozoa, which was not found in the serum of healthy dogs. The proteins are suspected to be disease markers, whereas the MALDI-TOF technique itself has high specificity and sensitivity and can be applied in analytical laboratories in the diagnosis of canine babesiosis.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)