Tobacco smoking-response genes in blood and buccal cells.
Toxicology letters (2014-12-03)
Hyun-Kyung Na, Minju Kim, Seong-Sil Chang, Soo-Young Kim, Jong Y Park, Myeon Woo Chung, Mihi Yang
ABSTRACT
Tobacco smoking is a well-known cause of various diseases, however, its toxic mechanisms for diseases are not completely understood, yet. Therefore, we performed biological monitoring to find tobacco smoking-responsive mechanisms including oxidative stress in Korean men (N=36). Whole genome microarray analyses were performed with peripheral blood from smokers and age-matched nonsmokers. We also performed qRT-PCR to confirm the microarray results and compared the gene expression of blood to those of buccal cells. To assess the effects of tobacco smoking on oxidative stress, we analyzed urinary levels of malondialdehyde (MDA), a lipid peroxidation marker, and performed PCR-based arrays on reactive oxygen species (ROS)-related genes. As results, 34 genes were differently expressed in blood between smokers and nonsmokers (ps<0.01 and >1.5-fold change). Particularly, the genes involved in immune responsive pathways, e.g., the Fcγ-receptor mediated phagocytosis and the leukocyte transendothelial migration pathways, were differentially expressed between smokers and nonsmokers. Among the above genes, the ACTG1, involved in the maintenance of actin cytoskeleton, cell migration and cancer metastasis, was highly expressed by smoking in both blood and buccal cells. Concerning oxidative stress, smokers showed high levels of urinary MDA and down-regulation of expressions of antioxidant related genes including TPO, MPO, GPX2, PTGR1, and NUDT1 as compared to nonsmokers (ps<0.05). In conclusion, these results suggest that systemically altered immune response and oxidative stress can be tobacco-responsive mechanisms for the related diseases. Based on consistent results in blood and buccal cells, expression of the ACTG1 can be a tobacco smoking-responsive biomarker.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ~98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Triethyl phosphate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%