  • Search Results
  • A column-switching HPLC-MS/MS method for mucopolysaccharidosis type I analysis in a multiplex assay for the simultaneous newborn screening of six lysosomal storage disorders.
A column-switching HPLC-MS/MS method for mucopolysaccharidosis type I analysis in a multiplex assay for the simultaneous newborn screening of six lysosomal storage disorders.
Biomedical chromatography : BMC (2014-01-23)
Antonina Gucciardi, Elisa Legnini, Iole Maria Di Gangi, Carlo Corbetta, Rosella Tomanin, Maurizio Scarpa, Giuseppe Giordano
ABSTRACT
Lysosomal storage disorders comprise a group of rare genetic diseases in which a deficit of specific hydrolases leads to the storage of undegraded substrates in lysosomes. Impaired enzyme activities can be assessed by MS/MS quantification of the reaction products obtained after incubation with specific substrates. In this study, a column-switching HPLC-MS/MS method for multiplex screening in dried blood spot of the lysosomal enzymes activities was developed. Mucopolysaccharidosis type I, Fabry, Gaucher, Krabbe, Niemann-Pick A/B and Pompe diseases were simultaneously assayed. Dried blood spots were incubated with substrates and internal standards; thereafter, supernatants were collected with minor manipulations. Samples were injected, trapped into an online perfusion column and, by a six-port valve, switched online through the C18 analytical column to perform separation of metabolites followed by MS/MS analysis. A total of 1136 de-identified newborn screening samples were analyzed to determine references for enzymes activity values. As positive controls, we analyzed dried blood spots from three patients with Pompe, one with Fabry, one with Krabbe disease and two with MPS I, and in all cases the enzyme activities were below the cutoff values measured for newborns, except for an MPS I patient after successful hematopoietic stem cell transplantation.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%