  • Search Results
  • Tissue-specific metabolite profiling of Turmeric by using laser micro-dissection, ultra-high performance liquid chromatography-quadrupole time of fight-mass spectrometry and liquid chromatography-tandem mass spectrometry.
Tissue-specific metabolite profiling of Turmeric by using laser micro-dissection, ultra-high performance liquid chromatography-quadrupole time of fight-mass spectrometry and liquid chromatography-tandem mass spectrometry.
European journal of mass spectrometry (Chichester, England) (2015-02-25)
Yogini Jaiswal, Zhitao Liang, Alan Ho, Hubiao Chen, Zhongzhen Zhao
ABSTRACT
Curcuma longa L. is recognized for its therapeutic and culinary uses both in Ayurveda and traditional Chinese medicine and is considered to be a boon to mankind. It has been extensively studied for its benefits and still continues to be an important drug with continued potential for further exploration and research. We studied the tissue-specific distribution of secondary metabolites to establish the validity of the use of rhizome samples from India and China, as substitutes for each other, based upon their metabolite profiles and curcumin contents. Laser microdissection was used for the isolation of microscopic tissues, such as cork, cortex and leaf-trace vascular bundles from rhizomes. Metabolite profiling was carried out by ultra-high performance liquid chromatography-quadrupole time of fight-mass spectrometry and curcumin content was estimated by a method validated as per the Harmonized Tripartite Guidelines. The cortex and cork revealed the presence of a higher number of secondary metabolites than in the leaf-trace vascular bundles. The curcumin contents in rhizome samples from both the countries, estimated with the help of a precise and accurate validated method, were found to be comparable. Based on the results, we conclude that turmeric rhizomes grown in India and China are qualitatively and quantitatively indistinguishable and therefore can be used as substitutes. The developed method can be widely applied for microscopic identification, authentication and analysis of the distribution of phytoconstituents in other botanical species of interest or of species with a significant commercial and therapeutic value.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)