  • Search Results
  • Resolvin D1 and lipoxin A4 improve alveolarization and normalize septal wall thickness in a neonatal murine model of hyperoxia-induced lung injury.
Resolvin D1 and lipoxin A4 improve alveolarization and normalize septal wall thickness in a neonatal murine model of hyperoxia-induced lung injury.
PloS one (2014-06-04)
Camilia R Martin, Munir M Zaman, Calvin Gilkey, Maria V Salguero, Hatice Hasturk, Alpdogan Kantarci, Thomas E Van Dyke, Steven D Freedman
ABSTRACT
The critical fatty acids Docosahexaenoic Acid (DHA) and Arachidonic Acid (AA) decline in preterm infants within the first postnatal week and are associated with neonatal morbidities, including bronchopulmonary dysplasia (BPD). DHA and AA are precursors to downstream metabolites that terminate the inflammatory response. We hypothesized that treatment with Resolvin D1 and/or Lipoxin A4 would prevent lung injury in a murine model of BPD. To determine the effect of Resolvin D1 and/or Lipoxin A4 on hyperoxia-induced lung injury. C57/BL6 pups were randomized at birth to Room Air, Hyperoxia (>90% oxygen), Hyperoxia + Resolvin D1, Hyperoxia + Lipoxin A4, or Hyperoxia + Resolvin D1/Lipoxin A4. Resolvin D1 and/or Lipoxin A4 (2 ng/g) were given IP on days 0, 3, 6, and 9. On day 10, mice were sacrificed and lungs collected for morphometric analyses including Mean Linear Intercept (MLI), Radial Alveolar Count (RAC), and Septal Thickness (ST); RT-PCR analyses of biomarkers of lung development and inflammation; and ELISA for TGFβ1 and TGFβ2. The increased ST observed with hyperoxia exposure was normalized by both Resolvin D1 and Lipoxin A4; while, hyperoxia-induced alveolar simplification was attenuated by Lipoxin A4. Relative to hyperoxia, Resolvin D1 reduced the gene expression of CXCL2 (2.9 fold), TIMP1 (6.7 fold), and PPARγ (4.8 fold). Treatment with Lipoxin A4 also led to a reduction of CXCL2 (2.4 fold) while selectively increasing TGFβ2 (2.1 fold) and Smad3 (1.58 fold). The histologic and biochemical changes seen in hyperoxia-induced lung injury in this murine model can be reversed by the addition of DHA and AA fatty acid downstream metabolites that terminate the inflammatory pathways and modulate growth factors. These fatty acids or their metabolites may be novel therapies to prevent or treat lung injury in preterm infants.
Product Description
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of Ph. Eur., BP, 35 wt. %, contains 10% methanol as stabilizer
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O