  • Search Results
  • High genotypic and phenotypic similarity among Shiga toxin-producing Escherichia coli O111 environmental and outbreak strains.
High genotypic and phenotypic similarity among Shiga toxin-producing Escherichia coli O111 environmental and outbreak strains.
Foodborne pathogens and disease (2015-02-07)
Michelle E Diodati, Anne H Bates, Michael B Cooley, Samarpita Walker, Robert E Mandrell, Maria T Brandl
ABSTRACT
Escherichia coli serogroup O111 is among the six most commonly reported non-O157:H7 Shiga toxin-producing E. coli (STEC), which are emerging as important foodborne pathogens. We have assembled a collection of environmental and clinical strains of E. coli O111 from diverse sources and investigated various genotypic and phenotypic characteristics of these strains to gain a better understanding of the epidemiology and biology of this serogroup. Sixty-three percent of the strains (24/38) were of H-type 8, which dominated the environmental- and outbreak-strains group, whereas the sporadic-case strains were more heterogeneous in H-type. All of the environmental and outbreak strains harbored the Shiga toxin 1 gene (stx1), eae, and ehx, and a subset of these also carried the Shiga toxin 2 gene (stx2). Only 9 of 16 sporadic-case strains produced stx1 and/or stx2, and these were mostly of H-type 8 and 10. Pulsed-field gel electrophoresis analysis revealed a cluster of environmental, outbreak, and sporadic illness strains with high phylogenetic similarity. Strains in this pulsogroup were all of the H8 type and STEC pathotype, and carried eae and ehx. Smaller clusters of highly similar STEC O111 strains included outbreak and sporadic illness strains isolated during different time periods or from different geographical locations. A distinct aggregative behavior was observed in the cultures of all environmental and outbreak STEC O111 strains, but not in those of sporadic-case strains. Among environmental and outbreaks strains, aggregation was positively correlated with production of curli fimbriae and RpoS function, and negatively with cellulose synthesis, while the nonaggregative behavior of sporadic-case strains correlated (positively) only with cellulose production. Our results indicate that STEC O111 strains sharing high genotypic similarity and important phenotypic traits with STEC O111 outbreak strains are present in the agricultural environment and may contribute to the burden of foodborne disease.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, Vetec, reagent grade, 99%
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid