  • Search Results
  • Isolation and Bioactivity Analysis of Ethyl Acetate Extract from Acer tegmentosum Using In Vitro Assay and On-Line Screening HPLC-ABTS(+) System.
Isolation and Bioactivity Analysis of Ethyl Acetate Extract from Acer tegmentosum Using In Vitro Assay and On-Line Screening HPLC-ABTS(+) System.
Journal of analytical methods in chemistry (2014-11-12)
Kwang Jin Lee, Na-Young Song, You Chang Oh, Won-Kyung Cho, Jin Yeul Ma
ABSTRACT
The Acer tegmentosum (3 kg) was extracted using hot water, and the freeze-dried extract powder was partitioned successively using dichloromethane (DCM), ethyl acetate (EA), butyl alcohol (n-BuOH), and water. From the EA extract fraction (1.24 g), five phenolic compounds were isolated by the silica gel, octadecyl silica gel, and Sephadex LH-20 column chromatography. Based on spectroscopic methods such as (1)H-NMR, (13)C-NMR, and LC/MS the chemical structures of the compounds were confirmed as feniculin (1), avicularin (2), (+)-catechin (3), (-)-epicatechin (4), and 6'-O-galloyl salidroside (5). Moreover, a rapid on-line screening HPLC-ABTS(+) system for individual bioactivity of the EA-soluble fraction (five phenolic compounds) was developed. The results indicated that compounds 1 and 2 were first isolated from the A. tegmentosum. The anti-inflammatory activities and on-line screening HPLC-ABTS(+) assay method of these compounds in LPS-stimulated murine macrophages were rapid and efficient for the investigation of bioactivity of A. tegmentosum.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)