  • Search Results
  • Genomic analysis of the interaction between pesticide exposure and nutrition in honey bees (Apis mellifera).
Genomic analysis of the interaction between pesticide exposure and nutrition in honey bees (Apis mellifera).
Journal of insect physiology (2014-12-03)
Daniel R Schmehl, Peter E A Teal, James L Frazier, Christina M Grozinger
ABSTRACT
Populations of pollinators are in decline worldwide. These declines are best documented in honey bees and are due to a combination of stressors. In particular, pesticides have been linked to decreased longevity and performance in honey bees; however, the molecular and physiological pathways mediating sensitivity and resistance to pesticides are not well characterized. We explored the impact of coumaphos and fluvalinate, the two most abundant and frequently detected pesticides in the hive, on genome-wide gene expression patterns of honey bee workers. We found significant changes in 1118 transcripts, including genes involved in detoxification, behavioral maturation, immunity, and nutrition. Since behavioral maturation is regulated by juvenile hormone III (JH), we examined effects of these miticides on hormone titers; while JH titers were unaffected, titers of methyl farnesoate (MF), the precursor to JH, were decreased. We further explored the association between nutrition- and pesticide-regulated gene expression patterns and demonstrated that bees fed a pollen-based diet exhibit reduced sensitivity to a third pesticide, chlorpyrifos. Finally, we demonstrated that expression levels of several of the putative pesticide detoxification genes identified in our study and previous studies are also upregulated in response to pollen feeding, suggesting that these pesticides and components in pollen modulate similar molecular response pathways. Our results demonstrate that pesticide exposure can substantially impact expression of genes involved in several core physiological pathways in honey bee workers. Additionally, there is substantial overlap in responses to pesticides and pollen-containing diets at the transcriptional level, and subsequent analyses demonstrated that pollen-based diets reduce workers' pesticide sensitivity. Thus, providing honey bees and other pollinators with high quality nutrition may improve resistance to pesticides.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Coumaphos, PESTANAL®, analytical standard
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)