  • Search Results
  • Ocular pharmacokinetics of naringenin eye drops following topical administration to rabbits.
Ocular pharmacokinetics of naringenin eye drops following topical administration to rabbits.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics (2014-09-18)
Junli Lin, Jingjing Sun, Yandong Wang, Yan Ma, Wenpei Chen, Ziyang Zhang, Gang Gui, Baoqin Lin
ABSTRACT
To investigate the ocular pharmacokinetics of 1% naringenin eye drops following topical administration to rabbits. One drop (50 μL) of 1% naringenin eye drops was instilled into both eyes of each rabbit. The animals were sacrificed at predetermined intervals after dosing, and ocular tissues and plasma were then collected. Concentrations of naringenin were analyzed using specific electrospray ionization liquid chromatography-tandem mass spectrometry method, which is proved to be sensitive, specific, precise, and suitable for determination of naringenin in ocular tissues and plasma of rabbits. Ocular exposure to naringenin, based on AUC(0-t), was highest in cornea, followed by aqueous humor, retina, and vitreous body. The Cmax of naringenin in cornea, aqueous humor, vitreous body, and retina were 67945.30 ± 4109.34 ng/g, 1325.69 ± 239.34, 160.52 ± 38.78 ng/mL, and 1927.08 ± 660.77 ng/g at 0.083, 0.75, 0.083, and 0.083 h after topical administration, respectively. The half-lives for these tissues were 9.37, 0.65, 1.17, and 4.62 h, respectively. There was no significant difference between free naringenin and total naringenin in plasma based on Cmax and Tmax. Cmax of total naringenin in plasma at 0.083 h was 35.12 ± 0.54 ng/mL. Measurable concentrations of naringenin were achieved in ocular tissues after topical application in rabbits. Topical instillation of naringenin may be an effective approach in the treatment of posterior section diseases.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Ammonium acetate, suitable for mass spectrometry (MS), LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Supelco
(±)-Naringenin, analytical standard
Sigma-Aldrich
Naringenin, natural (US), 98%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%