  • Search Results
  • Expression of antiviral cytokines in Crandell-Reese feline kidney cells pretreated with Korean red ginseng extract or ginsenosides.
Expression of antiviral cytokines in Crandell-Reese feline kidney cells pretreated with Korean red ginseng extract or ginsenosides.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-05-06)
Min Hwa Lee, Dong Joo Seo, Ju-Hee Kang, Seung Hyun Oh, Changsun Choi
ABSTRACT
The antiviral activity and protective mechanism of Korean red ginseng (KRG) is not well understood. The aim of this study was to investigate the protective mechanism of KRG extract and ginsenosides against feline calicivirus (FCV), a human norovirus surrogate. CRFK cells that were pretreated for 48h with 10μg/mL of KRG extract or purified ginsenoside Rb1 or Rg1, were inoculated with FCV. RNA extracted from each treated group was examined for the expression of antiviral cytokines, including interferon-α (IFN-α), interferon-β (IFN-β), interferon-ω (IFN-ω), Mx, and zinc finger antiviral protein shorter isoform (ZAPS), by relative real-time reverse transcription-polymerase chain reaction. mRNA expression of IFN-α, IFN-β, IFN-ω, Mx, and ZAPS was significantly induced in the FCV-challenged group pretreated with the KRG extract or ginsenosides, and it was higher than the group treated with FCV alone. Mx protein expression was confirmed by western blotting of CRFK cells pretreated with the ginsenoside Rb1 or with Rg1. Induction of antiviral cytokines contributes to the reduction of the viral titer in CRFK cells pretreated with the KRG extract and purified ginsenosides. In future studies, the antiviral protective mechanism of KRG should be demonstrated using other viruses such as human norovirus.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Interferon α 2A human, IFN-α 2A, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%