  • Search Results
  • Legionella dumoffii utilizes exogenous choline for phosphatidylcholine synthesis.
Legionella dumoffii utilizes exogenous choline for phosphatidylcholine synthesis.
International journal of molecular sciences (2014-05-14)
Marta Palusinska-Szysz, Agnieszka Szuster-Ciesielska, Magdalena Kania, Monika Janczarek, Elżbieta Chmiel, Witold Danikiewicz
ABSTRACT
Phosphatidycholine (PC) is the major membrane-forming phospholipid in eukaryotes but it has been found in only a limited number of prokaryotes. Bacteria synthesize PC via the phospholipid N-methylation pathway (Pmt) or via the phosphatidylcholine synthase pathway (Pcs) or both. Here, we demonstrated that Legionella dumoffii has the ability to utilize exogenous choline for phosphatidylcholine (PC) synthesis when bacteria grow in the presence of choline. The Pcs seems to be a primary pathway for synthesis of this phospholipid in L. dumoffii. Structurally different PC species were distributed in the outer and inner membranes. As shown by the LC/ESI-MS analyses, PC15:0/15:0, PC16:0/15:0, and PC17:0/17:1 were identified in the outer membrane and PC14:0/16:0, PC16:0/17:1, and PC20:0/15:0 in the inner membrane. L. dumoffii pcsA gene encoding phosphatidylcholine synthase revealed the highest sequence identity to pcsA of L. bozemanae (82%) and L. longbeachae (81%) and lower identity to pcsA of L. drancourtii (78%) and L. pneumophila (71%). The level of TNF-α in THP1-differentiated cells induced by live and temperature-killed L. dumoffii cultured on a medium supplemented with choline was assessed. Live L. dumoffii bacteria cultured on the choline-supplemented medium induced TNF-α three-fold less efficiently than cells grown on the non-supplemented medium. There is an evident effect of PC modification, which impairs the macrophage inflammatory response.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Supelco
Ammonium acetate, suitable for mass spectrometry (MS), LiChropur, eluent additive for LC-MS
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)