  • Search Results
  • A smart upconversion-based mesoporous silica nanotheranostic system for synergetic chemo-/radio-/photodynamic therapy and simultaneous MR/UCL imaging.
A smart upconversion-based mesoporous silica nanotheranostic system for synergetic chemo-/radio-/photodynamic therapy and simultaneous MR/UCL imaging.
Biomaterials (2014-08-12)
Wenpei Fan, Bo Shen, Wenbo Bu, Feng Chen, Qianjun He, Kuaile Zhao, Shengjian Zhang, Liangping Zhou, Weijun Peng, Qingfeng Xiao, Dalong Ni, Jianan Liu, Jianlin Shi
ABSTRACT
To achieve the accurate diagnosis and efficient in situ therapy of malignant tumors is encouraging but still remains a big challenge. The integration of upconversion nanoparticles and mesoporous silica that can combine the diagnostic/therapeutic functions within a single platform, may provide a more advanced way for the efficient theranostics of cancer. In this study, sub-80 nm rattle-structured multifunctional Gd-UCNPs core/mesoporous silica shell nanotheranostics (UCMSNs) were successfully constructed for the co-delivery of a radio-/photo-sensitizer hematoporphyrin (HP) and a radiosensitizer/chemodrug docetaxel (Dtxl). Upon NIR excitation and X-ray irradiation, the complete tumor elimination has been achieved by the synergetic chemo-/radio-/photodynamic tri-modal therapy under the assistance of simultaneous magnetic/upconversion luminescent (MR/UCL) bimodal imaging. To the best of our knowledge, this study is the first example of achieving tri-modal synergetic therapy in one single nanotheranostic system, and we anticipate that it may serve as a particularly useful platform for the clinical diagnosis and efficient treatment of cancer from bench to beside.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%