  • Search Results
  • Simultaneous determination of phentermine and topiramate in human plasma by liquid chromatography-tandem mass spectrometry with positive/negative ion-switching electrospray ionization and its application in pharmacokinetic study.
Simultaneous determination of phentermine and topiramate in human plasma by liquid chromatography-tandem mass spectrometry with positive/negative ion-switching electrospray ionization and its application in pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2015-02-11)
Yang Ni, Ying Zhou, Mingzhen Xu, Xiaomeng He, Huqun Li, Satter Haseeb, Hui Chen, Weiyong Li
ABSTRACT
A new method for simultaneous determination of phentermine and topiramate by liquid chromatography/electrospray tandem mass spectrometry (LC/MS/MS) operated in positive and negative ionization switching modes was developed and validated. Protein precipitation with acetonitrile was selected for sample preparation. Analyses were performed on a liquid chromatography system employing a Kromasil 60-5CN column (2.1 mm × 100 mm, 5 μm) and an isocratic elution with mixed solution of acetonitrile-20mM ammonium formate containing 0.3% formic acid (40:60, v/v), at a flow rate of 0.35 mL/min. Doxazosin mesylate and pioglitazone were used as the internal standard (IS) respectively for quantification. The determination was carried out on an API 4000 triple-quadrupole mass spectrometer operated in multiple reaction monitoring (MRM) mode using the following transitions monitored simultaneously: positive m/z 150.0/91.0 for phentermine, m/z 452.1/344.3 for doxazosin, and negative m/z 338.3/77.9 for topiramate, m/z 355.0/41.9 for pioglitazone. The method was validated to be linear over the concentration range of 1-800 ng mL(-1) for phentermine, 1-1000 ng mL(-1) for topiramate. Within- and between-day accuracy and precision of the validated method at three different concentration levels were within the acceptable limits of <15% at all concentrations. Blood samples were collected into heparinized tubes before and after administration. The simple and robust LC/MS/MS method was successfully applied for the simultaneous determination of phentermine and topiramate in a pharmacokinetic study in healthy male Chinese volunteers.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free