  • Search Results
  • Tailored polymer-lipid hybrid nanoparticles for the delivery of drug conjugate: dual strategy for brain targeting.
Tailored polymer-lipid hybrid nanoparticles for the delivery of drug conjugate: dual strategy for brain targeting.
Colloids and surfaces. B, Biointerfaces (2015-01-21)
Udita Agrawal, Gousia Chashoo, Parduman Raj Sharma, Ashok Kumar, Ajit Kumar Saxena, S P Vyas
ABSTRACT
The object of the present study was to investigate the glioma targeting propensity of folic acid (F) decorated polymer-lipid hybrid nanoparticles (PLNs) encapsulating cyclo-[Arg-Gly-Asp-D-Phe-Lys] (cRGDfK) modified paclitaxel (PtxR-FPLNs). The prepared PLNs were supposed to bypass the blood-brain barrier (BBB) efficiently and subsequently target integrin rich glioma cells. The developed formulations were characterized for size, shape, drug entrapment efficiency, and in vitro release profile. PtxR-FPLNs demonstrated highest in vitro inhibitory effect, cell apoptosis and cell uptake. Pharmacokinetics and biodistribution studies showed efficacy of PtxR-FPLNs in vivo. In vivo anti-tumor studies clearly revealed that the median survival time for Balb/C mice treated with PtxR-FPLNs (42 days) was extended significantly as compared to PtxR-PLNs (35 days), free PtxR (18 days), Ptx-FPLNs (38 days), Ptx-PLNs (30 days), free Ptx (14 days) and control group (12 days). From the results it can be concluded that the developed dual targeted nanoformulation was able to efficiently cross the BBB and significantly deliver higher amounts of drug to brain tumor for better therapeutic outcome.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Folic acid, meets USP testing specifications