  • Search Results
  • N-glycomic profiling of a glucosidase II mutant of Dictyostelium discoideum by ''off-line'' liquid chromatography and mass spectrometry.
N-glycomic profiling of a glucosidase II mutant of Dictyostelium discoideum by ''off-line'' liquid chromatography and mass spectrometry.
Electrophoresis (2014-02-28)
Alba Hykollari, Martin Dragosits, Dubravko Rendić, Iain B H Wilson, Katharina Paschinger
ABSTRACT
In this study, we have performed the first mass spectrometric analysis of N-glycans of the M31 mutant strain of the cellular slime mould Dictyostelium discoideum, previously shown to have a defect in glucosidase II. Together with glucosidase I, this enzyme mediates part of the initial processing of N-glycans; defects in either glucosidase are associated with human diseases and result in an accumulation of incorrectly processed oligosaccharides which are not, or only poor, substrates for a range of downstream enzymes. To examine the effect of the glucosidase II mutation in Dictyostelium, we employed off-line LC-MALDI-TOF MS in combination with chemical and enzymatic treatments and MS/MS to analyze the neutral and anionic N-glycans of the mutant as compared to the wild type. The major neutral species were, as expected, of the composition Hex10-11 HexNAc2-3 with one or two terminal glucose residues. Consistent with the block in processing of neutral N-glycans caused by the absence of glucosidase II, fucose was apparently absent from the N-glycans and bisecting N-acetylglucosamine was rare. The major anionic oligosaccharides were sulfated and/or methylphosphorylated forms of Hex8-11 HexNAc2-3 , many of which surprisingly lacked glucose residues entirely. As anionic N-glycans are considered to be mostly associated with lysosomal enzymes in Dictyostelium, we hypothesise that glycosidases present in the acidic compartments may act on the oligosaccharides attached to such slime mould proteins. Furthermore, our chosen analytical approach enabled us, via observation of diagnostic negative-mode MS/MS fragments, to determine the fine structure of the methylphosphorylated and sulfated N-glycans of the M31 glucosidase mutant in their native state.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl