  • Search Results
  • Role of Serotonin in MODS: Deficiency of Serotonin Protects Against Zymosan-Induced Multiple Organ Failure in Mice.
Role of Serotonin in MODS: Deficiency of Serotonin Protects Against Zymosan-Induced Multiple Organ Failure in Mice.
Shock (Augusta, Ga.) (2014-11-14)
Jingyao Zhang, Qing Pang, Sidong Song, Ruiyao Zhang, Sushun Liu, Zichao Huang, Qifei Wu, Yang Liu, Chang Liu
ABSTRACT
Zymosan-induced multiple organ dysfunction syndrome (MODS) is a multifactorial pathology that involves the deterioration of function of several organs. 5-Hydroxytryptamine (5-HT) is a small monoamine molecule that is primarily known for its role as a neurotransmitter. Previous studies have shown that 5-HT could serve as an important inflammatory mediator in the peripheral immune system. In the present study, we investigated the effect of 5-HT on the development of non-septic shock caused by zymosan in mice. Tryptophan hydroxylase 1-knockout mice (TPH1, leading to the absence of 5-HT), TPH1 + 5-hydroxytryptophan (precursor of 5-HT) treatment mice, wild-type (TPH1) mice, and wild-type plus p-chlorophenylalanine (PCPA, TPH1 inhibitor) treatment mice received zymosan intraperitoneally at a dose of 500 mg/kg. Organ failure and systemic inflammation in the mice were assessed 18 h after the administration of zymosan. Deficiency of 5-HT caused a significant reduction of the 1) peritoneal exudate formation, 2) neutrophil infiltration, 3) MODS, 4) nitrosative stress, and 5) cytokine formation. In addition, at the end of the observation period (7 days), deficiency of 5-HT in the mice was shown to be able to alleviate the severe illness characterized as systemic toxicity, significant loss of body weight, and high mortality caused by zymosan. In conclusion, the lack of 5-HT by genetic knockout or by pharmacologic inhibition of the TPH1 enzyme significantly attenuated zymosan-induced MODS.
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.5-100.5%
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material