  • Search Results
  • Involvement of Nod2 in the innate immune response elicited by malarial pigment hemozoin.
Involvement of Nod2 in the innate immune response elicited by malarial pigment hemozoin.
Microbes and infection (2014-12-03)
Yolanda Corbett, Silvia Parapini, Sarah D'Alessandro, Diletta Scaccabarozzi, Bruno C Rocha, Timothy J Egan, Aneesa Omar, Laura Galastri, Katherine A Fitzgerald, Douglas T Golenbock, Donatella Taramelli, Nicoletta Basilico
ABSTRACT
In malaria, the evidence concerning the nucleotide-binding, oligomerization domain (NOD) 2 (NOD2) receptor is fragmented and the stimuli that might activate NOD2 are not well characterized. We investigated the role of NOD2 in vitro in the response of macrophages to Plasmodium falciparum products. Immortalized or primary bone marrow derived macrophages from wild type C57Bl/6 mice, or knockout mice for NOD2 or its adaptor proteins, were either primed with interferon gamma or left untreated, and stimulated with parasite products. Both lysates of infected erythrocytes or hemozoin induced higher levels of nitric oxide in primed than in unprimed wild type macrophages. When stimulated with hemozoin, primed macrophages knockout for NOD2, or for its adaptor proteins, produced significantly lower nitric oxide levels compared to wild type cells. Differently from hemozoin, the use of β-hematin (synthetic hemozoin) as stimulus showed that NOD2 is dispensable. Furthermore, the production of inflammatory cytokines by wild type cells treated with hemozoin was not dependent on NOD2. These data indicate that parasite components present in the hemozoin, differently from β-hematin, induce the production of nitric oxide through the activation of NOD2, whereas the production of inflammatory cytokines, like TNF-α or MIP-2 (CXCL2), seems to be NOD2 independent.
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methylene blue, certified by the Biological Stain Commission
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine, Vetec, reagent grade, ≥99%
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)