  • Search Results
  • Carotenoids in durian fruit pulp during growth and postharvest ripening.
Carotenoids in durian fruit pulp during growth and postharvest ripening.
Food chemistry (2015-03-15)
Apinya Wisutiamonkul, Somnuk Promdang, Saichol Ketsa, Wouter G van Doorn
ABSTRACT
Durian (Durio zibethinus) cvs. Chanee and Monthong fruit were severed from the tree during 14 day intervals, from 10 weeks after anthesis until commercial maturity. We determined the pulp (i.e. aril; fruit flesh) carotenoid composition, together with pulp firmness, color and total soluble solids (TSS) and postharvest quality. In ripe cv. Chanee fruit the main carotenoids were β-carotene (about 80%), and α-carotene (20%), with minor levels of lutein and zeaxanthin. In ripe fruit total carotenoid concentration (expressed per gram FW) was about 9-fold higher in cv. Chanee than in cv. Monthong. Large differences between the cultivars were also found in β-carotene levels (about 11 times more in cv. Chanee), and even larger ones in those of α-carotene. Differences in lutein and zeaxanthin concentrations were small. Pulp color was deeper yellow in cv. Chanee than in cv. Monthong, which was correlated with α-carotene and β-carotene concentrations. Durian contains a high fat percentage, which is conducive to carotenoid uptake. It is concluded that it is advisable to consume cv. Chanee rather than cv. Monthong if intake of carotenoids is considered important.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Ammonium acetate, suitable for mass spectrometry (MS), LiChropur, eluent additive for LC-MS
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%