  • Search Results
  • Measurement of the direct oral anticoagulants apixaban, dabigatran, edoxaban, and rivaroxaban in human plasma using turbulent flow liquid chromatography with high-resolution mass spectrometry.
Measurement of the direct oral anticoagulants apixaban, dabigatran, edoxaban, and rivaroxaban in human plasma using turbulent flow liquid chromatography with high-resolution mass spectrometry.
Therapeutic drug monitoring (2014-04-04)
Tracey Gous, Lewis Couchman, Jignesh P Patel, Chitongo Paradzai, Roopen Arya, Robert J Flanagan
ABSTRACT
Direct oral anticoagulants (DOACs) are prescribed for systemic anticoagulation. Fixed doses are recommended, but dose individualization may be warranted. Functional coagulation assays may be available, but their use requires knowledge of the drug taken. To provide alternative methodology for guiding dosage, we have developed and validated a liquid chromatography-mass spectrometric assay for apixaban, dabigatran, edoxaban, and rivaroxaban at the concentrations attained during therapy. Samples, calibrators, and internal quality controls (100 μL) were mixed with internal standard solution (50 μg/L both dabigatran-13C6 and rivaroxaban-13C6 in acetonitrile) and, after centrifugation (16,400g, 4 minutes), supernatant (100 μL) was injected onto a Cyclone-C18-P-XL TurboFlow column. Analytes were focused onto an Accucore PhenylHexyl (2.1 × 100 mm, 2.6 μm) analytical column and eluted using a methanol + acetonitrile (1 + 1):aqueous ammonium acetate (10 mmol/L) gradient. Data were acquired using high-resolution mass spectrometry in full-scan mode (100-2000 m/z) with data-dependent fragmentation to confirm peak identity. Calibration was linear (1-500 μg/L all analytes). Total analysis time was 6 minutes. Intra-assay imprecision (% RSD) at 1 μg/L was 2.6%, 4.2%, 17.3%, and 9.5% for apixaban, dabigatran, edoxaban, and rivaroxaban, respectively. Mean recovery was 96%-101%. No signal suppression or enhancement was observed. Apixaban, dabigatran, and rivaroxaban were stable over 3 freeze-thaw cycles, after storage at room temperature, and at 2-8°C for up to 2 weeks. Edoxaban was stable over 3 freeze-thaw cycles but showed a mean deterioration of 16% if stored at 2-8°C (2 weeks) and of 18% and 70% (1 day and 2 weeks, respectively) at room temperature. The method is suitable for high-throughput therapeutic drug monitoring of DOACs. The acquisition of full scan data allows for the retrospective identification of metabolites. The method can be used to identify a particular DOAC if information on the drug taken is lacking.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Ammonium acetate, suitable for mass spectrometry (MS), LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)