  • Search Results
  • A pyridine-containing Cu²⁺-selective probe based on naphthalimide derivative.
A pyridine-containing Cu²⁺-selective probe based on naphthalimide derivative.
Sensors (Basel, Switzerland) (2014-12-18)
Jun Zhang, Qiang Wu, Bangliang Yu, Chunwei Yu
ABSTRACT
A new fluorescent probe P derived from naphthalimide bearing a pyridine group has been synthesized and characterized. The proposed probe P shows high selectivity and sensitivity to Cu2+ in aqueous media. Under optimized conditions, the linear response of P (2 μM) toward Cu2+ was 0.05-0.9 μM in ethanol-water solution (3:2, v:v, 50 mM HEPES, pH 7.4), and the detection limit was 0.03 μM.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D