  • Search Results
  • Determination of log P values of new cyclen based antimalarial drug leads using RP-HPLC.
Determination of log P values of new cyclen based antimalarial drug leads using RP-HPLC.
Die Pharmazie (2014-10-03)
A V Rudraraju, P N A Amoyaw, T J Hubin, M O F Khan
ABSTRACT
Lipophilicity, expressed by log P, is an important physicochemical property of drugs that affects many biological processes, including drug absorption and distribution. The main purpose of this study to determine the log P values of newly discovered drug leads using reversed-phase high-performance liquid chromatography (RP-HPLC). The reference standards, with varying polarity ranges, were dissolved in methanol and analyzed by RP-HPLC using a C18 column. The mobile phase consisted of a mixture of acetonitrile, methanol and water in a gradient elution mode. A calibration curve was plotted between the experimental log P values and obtained log k values of the reference standard compounds and a best fit line was obtained. The log k values of the new drug leads were determined in the same solvent system and were used to calculate the respective log P values by using the best fit equation. The log P vs. log k data gave a best fit linear curve that had an R2 of 0.9786 with Pvalues of the intercept and slope of 1.19 x 10(-6) and 1.56 x 10(-10), respectively, at 0.05 level of significance. Log P values of 15 new drug leads and related compounds, all of which are derivatives of macrocyclic polyamines and their metal complexes, were determined. The values obtained are closely related to the calculated log P (Clog P) values using ChemDraw Ultra 12.0. This experiment provided efficient, fast and reasonable estimates of log P values of the new drug leads by using RP-HPLC.
Sigma-Aldrich
Methanol, NMR reference standard
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), Customized plastic drum
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%