  • Search Results
  • Quantification of levornidazole and its metabolites in human plasma and urine by ultra-performance liquid chromatography-mass spectrometry.
Quantification of levornidazole and its metabolites in human plasma and urine by ultra-performance liquid chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-24)
Yuran Cao, Miao Zhao, Xiaojie Wu, Beining Guo, Yuancheng Chen, Jicheng Yu, Guoying Cao, Jing Zhang, Yaoguo Shi, Yingyuan Zhang
ABSTRACT
We developed and validated an ultra-performance liquid chromatographic (UPLC) method coupled with atmospheric pressure chemical ionization (APCI) mass spectrometry for simultaneous determination of levornidazole and its first-pass metabolites, l-chloro-3-(2-hydroxymethyl-5-nitro-l-imidazolyl)-2-propanol (Ml), 2-methyl-5-nitroimidazole (M2) and 3-(2-methyl-5-nitro-1-imidazolyl)-1,2-propanediol (M4), in human plasma and urine. The biological samples were pretreated by protein precipitation and liquid-liquid extraction and analyzed using an ACQUITY UPLC CSH C18 column (2.1×50 mm, 1.7 μm) and a QTRAP mass spectrometer in multiple reaction monitoring mode via APCI. Acetonitrile and 0.1% formic acid in water was used as the mobile phase in gradient elution at a flow rate of 0.6 mL/min. The lower limit of quantification of this method was 0.0100, 0.00500, 0.0200 and 0.00250 μg/mL for levornidazole, M1, M2 and M4, respectively. The linear calibration curves were obtained for levornidazole, M1, M2, and M4 over the range of 0.0100-5.00, 0.00500-2.50, 0.0200-10.0 and 0.00250-1.25 μg/mL, respectively. The intra- and inter-batch precision was less than 12.2% in plasma and less than 10.8% in urine. The intra- and inter-batch accuracy was 87.8-105.7% in plasma and 92.8-109.2% in urine. The mean recovery of levornidazole, M1, M2 and M4 was 91.1-105.1%, 95.8-103.8%, 87.8-96.8%, 96.8-100.6% from plasma and 96.0-100.9%, 96.9-107.9%, 95.1-102.7%, 103.7-105.9% from urine respectively. This method was validated under various conditions, including room temperature, freeze-thaw cycles, long-term storage at -40 ± 5°C, after pretreatment in the autosampler (at 10°C), and 10- and 100-fold dilution. This newly established analytical method was successfully applied in a pharmacokinetic study following single intravenous infusion of levornidazole in 24 healthy Chinese subjects.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%