  • Search Results
  • Graphene-derivatized silica as an efficient solid-phase extraction sorbent for pre-concentration of fluoroquinolones from water followed by liquid-chromatography fluorescence detection.
Graphene-derivatized silica as an efficient solid-phase extraction sorbent for pre-concentration of fluoroquinolones from water followed by liquid-chromatography fluorescence detection.
Journal of chromatography. A (2015-01-07)
Andrea Speltini, Michela Sturini, Federica Maraschi, Luana Consoli, Alberto Zeffiro, Antonella Profumo
ABSTRACT
This work presents a novel analytical method based on graphene for the determination of five widely used fluoroquinolones (FQs) in aqueous matrices. The procedure entails solid-phase extraction (SPE) on graphene-derivatized silica (200mg), followed by liquid chromatography with fluorescence detection. Monolayer graphene oxide (GO) flakes were covalently bonded onto aminopropyl silica microparticles, and then treated with aqueous hydrazine to obtain the reduced GO (RGO). The final material (RGO-silica) was characterized by thermogravimetric analysis (TGA), scanning electron microscopy (SEM) and BET analysis, and for the first time evaluated as mixed-mode sorbent for the SPE of FQs from natural waters. Accuracy was studied on tap/raw river water in the concentration range 5-10,000ngL(-1), obtaining mean absolute recoveries from 72 to 118%. The inter-day precision was good, showing relative standard deviations (RSDs) in the range 5-15%. Sample volumes up to 1L provided enrichment factors up to 1000, achieving accurate quantification of concentrations as low as 5ngL(-1). The analytes were simultaneously and quantitatively eluted from the RGO-silica cartridge in a single fraction by using acetonitrile combined with aqueous tetrabutyl ammonium hydroxide. The batch-to-batch reproducibility was verified on three independently prepared RGO-silica samples. RGO-silica was advantageous in terms of adsorption capacity and reusability with respect to commercial sorbents; the cartridge proved to be reusable for at least 10 consecutive extractions, with no significant loss of efficiency (recovery >70%). The analytical procedure was applied to the determination of FQs in actual environmental waters.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets