  • Search Results
  • Endogenous expression pattern of resolvin D1 in a rat model of self-resolution of lipopolysaccharide-induced acute respiratory distress syndrome and inflammation.
Endogenous expression pattern of resolvin D1 in a rat model of self-resolution of lipopolysaccharide-induced acute respiratory distress syndrome and inflammation.
International immunopharmacology (2014-09-15)
Wei Sun, Zai-ping Wang, Ping Gui, Weiyi Xia, Zhengyuan Xia, Xing-cai Zhang, Qing-zhu Deng, Wei Xuan, Christelle Marie, Lin-lin Wang, Qing-ping Wu, Tingting Wang, Yun Lin
ABSTRACT
Resolvin D1 (RvD1), an endogenous lipid mediator derived from docosahexaenoic acid, has been reported to promote a biphasic activity in anti-inflammatory response and regulate inflammatory resolution. The present study aimed to determine the endogenous expression pattern of RvD1 in a rat model of self-resolution of lipopolysaccharide (LPS)-induced acute respiratory distress syndrome (ARDS) and inflammation. The ARDS model was induced by administrating LPS (2mg/kg) via tracheotomy in 138 male Sprague-Dawley rats. At specified time points, lung injury and inflammation were respectively assessed by lung histology and analysis of bronchoalveolar lavage fluid and cytokine levels. The expression of endogenous RvD1 was detected by high performance liquid chromatography and tandem mass spectrometry. The results showed that histological lung injury peaked between 6h (LPS6h) and day 3, followed by recovery over 4-10 days after LPS administration. Lung tissue polymorph nuclear cell (PMN) was significantly increased at LPS6h, and peaked between 6h to day 2. The levels of interleukin (IL)-6 and IL-10 were significantly increased at LPS6h and remained higher over day 10 as compared to baseline. Intriguingly, the endogenous RvD1 expression was decreased gradually during the first 3 days, followed by almost completely recovery over days 9-10. The finding indicated that endogenous RvD1 underwent a decrease in expression followed by gradual increase that was basically coincident with the lung injury recovery in a rat model of self-resolution LPS-induced ARDS and inflammation. Our results may help define the optimal therapeutic window for endogenous RvD1 to prevent or treat LPS-induced ARDS and inflammation.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Supelco
Hexane, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol