  • Search Results
  • Optimization of diarylthiazole B-raf inhibitors: identification of a compound endowed with high oral antitumor activity, mitigated hERG inhibition, and low paradoxical effect.
Optimization of diarylthiazole B-raf inhibitors: identification of a compound endowed with high oral antitumor activity, mitigated hERG inhibition, and low paradoxical effect.
ChemMedChem (2014-11-29)
Maurizio Pulici, Gabriella Traquandi, Chiara Marchionni, Michele Modugno, Rosita Lupi, Nadia Amboldi, Elena Casale, Nicoletta Colombo, Luca Corti, Marina Fasolini, Fabio Gasparri, Wilma Pastori, Alessandra Scolaro, Daniele Donati, Eduard Felder, Arturo Galvani, Antonella Isacchi, Enrico Pesenti, Marina Ciomei
ABSTRACT
Aberrant activation of the mitogen-activated protein kinase (MAPK)-mediated pathway components, RAF-MEK-ERK, is frequently observed in human cancers and clearly contributes to oncogenesis. As part of a project aimed at finding inhibitors of B-Raf, a key player in the MAPK cascade, we originally identified a thiazole derivative endowed with high potency and selectivity, optimal in vitro ADME properties, and good pharmacokinetic profiles in rodents, but that suffers from elevated hERG inhibitory activity. An optimization program was thus undertaken, focused mainly on the elaboration of the R(1) and R(2) groups of the scaffold. This effort ultimately led to N-(4-{2-(1-cyclopropylpiperidin-4-yl)-4-[3-(2,5-difluorobenzenesulfonylamino)-2-fluorophenyl]thiazol-5-yl}-pyridin-2-yl)acetamide (20), which maintains favorable in vitro and in vivo properties, but lacks hERG liability. Besides exhibiting potent antiproliferative activity against only cell lines bearing B-Raf V600E or V600D mutations, compound 20 also intriguingly shows a weaker "paradoxical" activation of MEK in non-mutant B-Raf cells than other known B-Raf inhibitors. It also demonstrates very good efficacy in vivo against the A375 xenograft melanoma model (tumor volume inhibition >90% at 10 mg kg(-1) ); it is therefore a suitable candidate for preclinical development.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis