  • Search Results
  • Gastric cancer in three relatives of a patient with a biallelic IL12RB1 mutation.
Gastric cancer in three relatives of a patient with a biallelic IL12RB1 mutation.
Familial cancer (2014-12-04)
Ingrid P Vogelaar, Rachel S van der Post, Esther van de Vosse, J Han J M van Krieken, Nicoline Hoogerbrugge, Marjolijn J L Ligtenberg, Encarna Gómez García
ABSTRACT
IL-12Rβ1 deficiency, also known as immunodeficiency 30 (IMD30, OMIM 614891), is a rare immunodeficiency syndrome caused by biallelic mutations in IL12RB1. Three second-degree relatives of a patient with this syndrome, all women, developed intestinal-type gastric cancer (GC). In the Netherlands the incidence of non-cardia GC in women is only 7 per 100,000 person years. Both relatives that were available for testing proved to be heterozygous for the familial IL12RB1 mutation, suggesting there might be a causal relation. Testing 29 index patients from families with early onset and/or a familial history of GC for germline mutations in both IL12RB1 and IL12RB2, that encodes the binding partner of IL-12Rβ1, did not reveal other germline mutations in these genes. Therefore heterozygous inactivating mutations in IL12RB1 and IL12RB2 are unlikely to be frequently involved in GC predisposition. Additional research in families with IL12RB1 mutations is required to determine whether carriers of IL12RB1 mutations have an increased (gastric) cancer risk.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%