  • Search Results
  • Development and application of a robust N-glycan profiling method for heightened characterization of monoclonal antibodies and related glycoproteins.
Development and application of a robust N-glycan profiling method for heightened characterization of monoclonal antibodies and related glycoproteins.
Journal of pharmaceutical sciences (2014-05-21)
Tanya Q Shang, Andrew Saati, Kelly N Toler, Jianming Mo, Heyi Li, Tonya Matlosz, Xi Lin, Jennifer Schenk, Chee-Keng Ng, Toni Duffy, Thomas J Porter, Jason C Rouse
ABSTRACT
A highly robust hydrophilic interaction liquid chromatography (HILIC) method that involves both fluorescence and mass spectrometric detection was developed for profiling and characterizing enzymatically released and 2-aminobenzamide (2-AB)-derivatized mAb N-glycans. Online HILIC/mass spectrometry (MS) with a quadrupole time-of-flight mass spectrometer provides accurate mass identifications of the separated, 2-AB-labeled N-glycans. The method features a high-resolution, low-shedding HILIC column with acetonitrile and water-based mobile phases containing trifluoroacetic acid (TFA) as a modifier. This column and solvent system ensures the combination of robust chromatographic performance and full compatibility and sensitivity with online MS in addition to the baseline separation of all typical mAb N-glycans. The use of TFA provided distinct advantages over conventional ammonium formate as a mobile phase additive, such as, optimal elution order for sialylated N-glycans, reproducible chromatographic profiles, and matching total ion current chromatograms, as well as minimal signal splitting, analyte adduction, and fragmentation during HILIC/MS, maximizing sensitivity for trace-level species. The robustness and selectivity of HILIC for N-glycan analyses allowed for method qualification. The method is suitable for bioprocess development activities, heightened characterization, and clinical drug substance release. Application of this HILIC/MS method to the detailed characterization of a marketed therapeutic mAb, Rituxan(®), is described.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%