  • Search Results
  • Characterisation of multiple regulatory domains spanning the major transcriptional start site of the FUS gene, a candidate gene for motor neurone disease.
Characterisation of multiple regulatory domains spanning the major transcriptional start site of the FUS gene, a candidate gene for motor neurone disease.
Brain research (2014-12-03)
Kejhal Khursheed, Thomas P Wilm, Christine Cashman, John P Quinn, Vivien J Bubb, Diana J Moss
ABSTRACT
Fused-In-Sarcoma (FUS) is a candidate gene for neurological disorders including motor neurone disease and Parkinson׳s disease in addition to various types of cancer. Recently it has been reported that over expression of FUS causes motor neurone disease in mouse models hence mutations leading to changes in gene expression may contribute to the development of neurodegenerative disease. Genome evolutionary conservation was used to predict important cis-acting DNA regulators of the FUS gene promoter that direct transcription. The putative regulators identified were analysed in reporter gene assays in cells and in chick embryos. Our analysis indicated in addition to regulatory domains 5' of the transcriptional start site an important regulatory domain resides in intron 1 of the gene itself. This intronic domain functioned both in cell lines and in vivo in the neural tube of the chick embryo including developing motor neurones. Our data suggest the interaction of multiple domains including intronic domains are involved in expression of FUS. A better understanding of the regulation of expression of FUS may give insight into how its stimulus inducible expression may be associated with neurological disorders.
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
SAFC
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine