  • Search Results
  • Extreme urinary betaine losses in type 2 diabetes combined with bezafibrate treatment are associated with losses of dimethylglycine and choline but not with increased losses of other osmolytes.
Extreme urinary betaine losses in type 2 diabetes combined with bezafibrate treatment are associated with losses of dimethylglycine and choline but not with increased losses of other osmolytes.
Cardiovascular drugs and therapy (2014-07-26)
Michael Lever, Christopher J McEntyre, Peter M George, Sandy Slow, Jane L Elmslie, Helen Lunt, Stephen T Chambers, Amber Parry-Strong, Jeremy D Krebs
ABSTRACT
Betaine deficiency is a probable cardiovascular risk factor and a cause of elevated homocysteine. Urinary betaine excretion is increased by fibrate treatment, and is also often elevated in diabetes. Does fibrate further increase betaine excretion in diabetes, and does it affect the plasma concentrations and excretions of related metabolites and of other osmolytes? Samples from a previous study of type 2 diabetes were selected if participants were taking bezafibrate (n = 32). These samples were compared with participants matched for age and gender and not on a fibrate (comparator group, n = 64). Betaine, related metabolites, and osmolytes were measured in plasma and urine samples from these 96 participants. Median urinary betaine excretion in those on bezafibrate was 5-fold higher than in the comparator group (p < 0.001), itself 3.5-fold higher than the median reported for healthy populations. In the bezafibrate group, median dimethylglycine excretion was higher (9-fold, p < 0.001). Excretions of choline, and of the osmolytes myo-inositol, taurine and glycerophosphorylcholine, were not significantly different between groups. Some participants excreted more betaine than usual dietary intakes. Several betaine fractional clearances were >100 %. Betaine excretion correlated with excretions of the osmolytes myo-inositol and glycerophosphorylcholine, and also with the excretion of choline and N,N-dimethylglycine, but it was inconclusive whether these relationships were affected by bezafibrate therapy. Increased urinary betaine excretions in type 2 diabetes are further increased by fibrate treatment, sometimes to more than their dietary intake. Concurrent betaine supplementation may be beneficial.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)