  • Search Results
  • Determination of N-acetylglucosamine in cosmetic formulations and skin test samples by hydrophilic interaction liquid chromatography and UV detection.
Determination of N-acetylglucosamine in cosmetic formulations and skin test samples by hydrophilic interaction liquid chromatography and UV detection.
Journal of pharmaceutical and biomedical analysis (2015-01-16)
Alice Pedrali, Mariella Bleve, Priscilla Capra, Tobias Jonsson, Gabriella Massolini, Paola Perugini, Giorgio Marrubini
ABSTRACT
N-Acetylglucosamine is an ingredient in pharmaceuticals, nutritional supplements and in cosmetics. N-Acetylglucosamine in cosmetics is expected to improve skin hydration, reparation, and to contribute as anti-wrinkle agent. This study reports on the validation and application of an HPLC method based on HILIC and UV detection for determining N-acetylglucosamine in cosmetics and in samples obtained after testing the skin exposed to cosmetics formulations. The chromatographic column used is a ZIC(®)-pHILIC (150 mm × 4.6 mm, 5 μm particle size) on which a mobile phase containing acetonitrile-aqueous KH2PO4 (70:30, v/v) 15 mM was applied in isocratic elution mode injecting 20 μl of sample at 0.5 ml/min constant flow-rate and 10±1°C column temperature. Under these conditions the total run time was 10 min and N-acetylglucosamine eluted baseline separated from all other compounds in the samples. Calibration in the range from 40 to 80 μg/ml allowed to assess the method linearity (R(2)>0.999) in a concentration range corresponding to about 50% to 120% of the expected levels of N-acetylglucosamine in the formulations. Precision expressed by RSD% was always better than 2% in intra-day and inter-day assays of authentic samples. Accuracy was in all cases within 95-105% of the expected concentration value in formulations containing N-acetylglucosamine. The sensitivity of the method was at the level of 10 μg/ml as limit of detection, and at 40 μg/ml as limit of quantitation. The application of the method to formulations containing solid lipid nanoparticles documents its usefulness in cosmetic quality control. The results witness that the method is also suitable for the determination of N-acetylglucosamine in samples obtained from skin test strips.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetone, purum, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)