  • Search Results
  • Target genes involved in antiproliferative effect of modified ginseng extracts in lung cancer A549 cells.
Target genes involved in antiproliferative effect of modified ginseng extracts in lung cancer A549 cells.
Acta biochimica et biophysica Sinica (2014-04-20)
Keun-Hong Kim, Ilsan Choi, Yeon-Weol Lee, Chong-Kwan Cho, Hwa-Seung Yoo, Seung-Bae Lee, Suk Ho Choi, Ki-Rok Kwon, Jun-Hyeog Jang
ABSTRACT
Lung cancer is the most common cancer and the leading cause of cancer-related deaths. Panax ginseng has long been used to treat cancer and other diseases worldwide. Most of the pharmacological actions of ginseng are attributed to a variety of ginsenosides, which are often metabolized by intestinal bacteria into more effective forms. In this study, we found that the antiproliferative activity of ginseng was increased after enzymatic processing of ginseng saponin (50% inhibitory concentration, >70 μg/ml). To elucidate the mechanism by which modified ginseng extract (MGX) induced cell death in human lung cancer cells, the gene expression profiles of A549 cells regulated by MGX were assayed using Agilent PrimeView Human Gene Expression Arrays. The expression of 17 genes involved in the regulation of cell signaling, cell metabolism, transport, and cytoskeleton-regulation was up-regulated, whereas the expression of 16 genes implicated in invasion and metastasis and cellular metabolism was down-regulated in MGX-treated A549 cells. Moreover, nuclear staining with 4',6-diamidino-2-phenylindole revealed that MGX clearly caused nuclear condensation and fragmentation which are observed in apoptosis cell. These results elucidate crucial anticancer mechanisms of MGX and provide potential new targets for the assessment of anticancer activity of MGX.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)