  • Search Results
  • The impact of weakly bound ⁸⁹Zr on preclinical studies: non-specific accumulation in solid tumors and aspergillus infection.
The impact of weakly bound ⁸⁹Zr on preclinical studies: non-specific accumulation in solid tumors and aspergillus infection.
Nuclear medicine and biology (2015-01-15)
Gregory W Severin, Jesper T Jørgensen, Stefan Wiehr, Anna-Maria Rolle, Anders E Hansen, Andreas Maurer, Mike Hasenberg, Bernd Pichler, Andreas Kjær, Andreas I Jensen
ABSTRACT
Preclinical studies involving (89)Zr often report significant bone accumulation, which is associated with dissociation of the radiometal from the tracer. However, experiments determining the uptake of unbound (89)Zr in disease models are not performed as routine controls. The purpose of the present study was to investigate the impact of free or weakly bound (89)Zr on PET quantifications in disease models, in order to determine if such control experiments are warranted. Chemical studies were carried out to find a (89)Zr compound that would solubilize the (89)Zr as a weak chelate, thus mimicking free or weakly bound (89)Zr released in circulation. (89)Zr oxalate had the desired characteristics, and was injected into mice bearing FaDu and HT29 solid tumor xenografts, and mice infected in the lungs with the mold Aspergillus fumigatus, as well as in healthy controls (naïve). PET/CT or PET/MR imaging followed to quantify the distribution of the radionuclide in the disease models. (89)Zr oxalate was found to have a plasma half-life of 5.1 ± 2.3 h, accumulating mainly in the bones of all animals. Both tumor types accumulated (89)Zr on the order of 2-4 %ID/cm(3), which is comparable to EPR-mediated accumulation of certain species. In the aspergillosis model, the concentration of (89)Zr in lung tissue of the naïve animals was 6.0 ± 1.1 %ID/g. This was significantly different from that of the animals with advanced disease, showing 11.6 ± 1.8 %ID/g. Given the high levels of (89)Zr accumulation in the disease sites in the present study, we recommend control experiments mapping the biodistribution of free (89)Zr in any preclinical study employing (89)Zr where bone uptake is observed. Aqueous (89)Zr oxalate appears to be a suitable compound for such studies. This is especially relevant in studies where the tracer accumulation is based upon passive targeting, such as EPR.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Tantalum(V) ethoxide, packaged for use in deposition systems
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Supelco
L-Glutamine, certified reference material, TraceCERT®
Germanium, disks, 15mm, thickness 1.0mm, polycrystalline, 100%
Germanium, sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, sheet, 25x25mm, thickness 3.0mm, polycrystalline, 99.999%
Germanium, rod, 6mm, diameter 6.0mm, single crystal, 100%
Germanium, sheet, 50x50mm, thickness 3.0mm, polycrystalline, 99.999%
Germanium, sheet, 7x24mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, disks, 20mm, thickness 1.0mm, single crystal, 100%
Germanium, microfoil, 25x25mm, thinness 0.5μm, specific density 333μg/cm2, 6 micron aluminum permanent support, 100%
Germanium, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Germanium, sheet, 10x10mm, thickness 0.25mm, polycrystalline, 99.999%
Germanium, microfoil, 25x25mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Germanium, sheet, 25x25mm, thickness 1.0mm, single crystal, 99.999%
Germanium, rod, 25mm, diameter 5mm, polycrystalline, n-type, 99.999%
Germanium, sheet, 10x10mm, thickness 0.6mm, single crystal, -111, 100%
Germanium, sheet, 4x4mm, thickness 0.05mm, polycrystalline, 99.999%