  • Search Results
  • The separation and detection of PET tracers via capillary electrophoresis for chemical identity and purity analysis.
The separation and detection of PET tracers via capillary electrophoresis for chemical identity and purity analysis.
Journal of pharmaceutical and biomedical analysis (2014-02-19)
Shilin Cheung, Jimmy Ly, Mark Lazari, Saman Sadeghi, Pei Yuin Keng, R Michael van Dam
ABSTRACT
CE coupled with UV detection was assessed as a possible platform for the chemical identity and purity analysis of positron emission tomography (PET) tracers using [(18)F]FAC and [(18)F]FLT as examples. Representative samples containing mixtures of the tracers plus well-known impurities, as well as real radioactive samples (formulated for injection), were analyzed. Using MEKC with SDS in a neutral phosphate buffer, the separation of all compounds in the samples was achieved with baseline resolutions in less than 4.5min and 3min for FLT and FAC samples, respectively. In comparison to the gold-standard for chemical analysis (i.e. HPLC/UV), we have demonstrated improvements in analysis times, and comparable LOD. Although the reproducibility in migration time is slightly lower than that of the HPLC, identification of the compounds was still possible due to good peak separation. In addition, we show that CE can be used to identify and quantify Krytofix2.2.2 (a toxic and commonly used phase transfer catalyst) in less than 2min and with a LOD of 45μg/mL (non-optimized). These results demonstrate adequate performance for chemical identity and purity analysis. Combined with the potential for miniaturization into a microchip format, these results suggest the potential of CE as an integral part of a miniaturized quality control system for PET tracers.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Furfuryl alcohol, 98%
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC