  • Search Results
  • Depletion of penicillin G residues in heavy sows after intramuscular injection. Part I: tissue residue depletion.
Depletion of penicillin G residues in heavy sows after intramuscular injection. Part I: tissue residue depletion.
Journal of agricultural and food chemistry (2014-07-16)
S J Lupton, W L Shelver, D J Newman, S Larsen, D J Smith
ABSTRACT
Heavy sows (n = 126) were treated with penicillin G procaine at a 5× label dose (33 000 IU/kg) for 3 consecutive days by intramuscular (IM) injection using three patterns of drug administration. Treatments differed by injection pattern and injection volume. Sets of sows were slaughtered 5, 10, 15, 20, 25, 32, and 39 days after the last treatment; skeletal muscle, kidney, serum, and urine were collected for penicillin G analysis by LC-MS/MS. Penicillin G at withdrawal day 5 averaged 23.5 ± 10.5 and 3762 ± 1932 ng/g in muscle and kidney, respectively. After 15 days of withdrawal, muscle penicillin G residues were quantifiable in only one treated hog (3.4 ng/g) but averaged 119 ± 199 ng/g in kidneys. Using a hypothetical tolerance of 50 ng/g and a natural log-linear depletion model, the withdrawal period required for penicillin depletion to 50 ng/g was 11 days for skeletal muscle and 47 days for kidney.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Ammonium acetate, suitable for mass spectrometry (MS), LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard