  • Search Results
  • Microwave-assisted derivatization: application to steroid profiling by gas chromatography/mass spectrometry.
Microwave-assisted derivatization: application to steroid profiling by gas chromatography/mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-05-02)
Gregori Casals, Josep Marcos, Oscar J Pozo, José Alcaraz, María Jesús Martínez de Osaba, Wladimiro Jiménez
ABSTRACT
Gas chromatography-mass spectrometry (GC-MS) remains as the gold-standard technique for the study of the steroid metabolome. A main limitation is the need of performing a derivatization step since incubation with strong silylations agents for long periods of time (usually 16 h) is required for the derivatization of hindered hydroxyls present in some steroids of interest. In the present work, a rapid, simple and reproducible microwave-assisted derivatization method was developed. In the method, 36 steroids already treated with methoxyamine (2% in pyridine) were silylated with 50 μl of N-trimethylsilylimidazole by using microwave irradiation, and the formed methyloxime-trimethylsilyl derivatives were analyzed by GC-MS. Microwave power and derivatization time silylation conditions were optimized being the optimum conditions 600 W and 3 min respectively. In order to evaluate the usefulness of this technique, the urine steroid profiles for 20 healthy individuals were analyzed. The results of a comparison of microwave irradiation with the classical heating protocol showed similar derivatization yields, thus suggesting that microwave-assisted silylation is a valid tool for the rapid steroid metabolome study.
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Cyclohexane, Laboratory Reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methanol, Absolute - Acetone free