  • Search Results
  • Identification of a new reactive metabolite of pyrrolizidine alkaloid retrorsine: (3H-pyrrolizin-7-yl)methanol.
Identification of a new reactive metabolite of pyrrolizidine alkaloid retrorsine: (3H-pyrrolizin-7-yl)methanol.
Chemical research in toxicology (2014-10-09)
Muluneh M Fashe, Risto O Juvonen, Aleksanteri Petsalo, Minna Rahnasto-Rilla, Seppo Auriola, Pasi Soininen, Jouko Vepsäläinen, Markku Pasanen
ABSTRACT
Pyrrolizidine alkaloids (PAs) such as retrorsine are common food contaminants that are known to be bioactivated by cytochrome P450 enzymes to putative hepatotoxic, genotoxic, and carcinogenic metabolites known as dehydropyrrolizidine alkaloids (DHPs). We compared how both electrochemical (EC) and human liver microsomal (HLM) oxidation of retrorsine could produce short-lived intermediate metabolites; we also characterized a toxicologically important metabolite, (3H-pyrrolizin-7-yl)methanol. The EC cell was coupled online or offline to a liquid chromatograph/mass spectrometer (LC/MS), whereas the HLM oxidation was performed in 100 mM potassium phosphate (pH 7.4) in the presence of NADPH at 37 °C. The EC cell oxidation of retrorsine produced 12 metabolites, including dehydroretrorsine (m/z 350, [M + H(+)]), which was degraded to a new reactive metabolite at m/z 136 ([M + H(+)]). The molecular structure of this small metabolite was determined using high-resolution mass spectrometry and NMR spectroscopy followed by chemical synthesis. In addition, we also identified another minor but reactive metabolite at m/z 136, an isomer of (3H-pyrrolizin-7-yl)methanol. Both (3H-pyrrolizin-7-yl)methanol and its minor isomer were also observed after HLM oxidation of retrorsine and other hepatotoxic PAs such as lasiocarpine and senkirkin. In the presence of reduced glutathione (GSH), each isomer formed identical GSH conjugates at m/z 441 and m/z 730 in the negative ESI-MS. Because (3H-pyrrolizine-7-yl)methanol) and its minor isomer subsequently reacted with GSH, it is concluded that (3H-pyrrolizin-7-yl)methanol may be a common toxic metabolite arising from PAs.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%