  • Search Results
  • DNA binding and condensation properties of the herpes simplex virus type 1 triplex protein VP19C.
DNA binding and condensation properties of the herpes simplex virus type 1 triplex protein VP19C.
PloS one (2014-08-15)
Alakesh Bera, Edward M Perkins, Jian Zhu, Heng Zhu, Prashant Desai
ABSTRACT
Herpesvirus capsids are regular icosahedrons with a diameter of a 125 nm and are made up of 162 capsomeres arranged on a T = 16 lattice. The capsomeres (VP5) interact with the triplex structure, which is a unique structural feature of herpesvirus capsid shells. The triplex is a heterotrimeric complex; one molecule of VP19C and two of VP23 form a three-pronged structure that acts to stabilize the capsid shell through interactions with adjacent capsomeres. VP19C interacts with VP23 and with the major capsid protein VP5 and is required for the nuclear localization of VP23. Mutation of VP19C results in the abrogation of capsid shell synthesis. Analysis of the sequence of VP19C showed the N-terminus of VP19C is very basic and glycine rich. It was hypothesized that this domain could potentially bind to DNA. In this study an electrophoretic mobility shift assay (EMSA) and a DNA condensation assay were performed to demonstrate that VP19C can bind DNA. Purified VP19C was able to bind to both a DNA fragment of HSV-1 origin as well as a bacterial plasmid sequence indicating that this activity is non-specific. Ultra-structural imaging of the nucleo-protein complexes revealed that VP19C condensed the DNA and forms toroidal DNA structures. Both the DNA binding and condensing properties of VP19C were mapped to the N-terminal 72 amino acids of the protein. Mutational studies revealed that the positively charged arginine residues in this N-terminal domain are required for this binding. This DNA binding activity, which resides in a non-conserved region of the protein could be required for stabilization of HSV-1 DNA association in the capsid shell.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG