  • Search Results
  • Occurrence and behaviour of 105 active pharmaceutical ingredients in sewage waters of a municipal sewer collection system.
Occurrence and behaviour of 105 active pharmaceutical ingredients in sewage waters of a municipal sewer collection system.
Water research (2014-04-29)
Richard H Lindberg, Marcus Östman, Ulrika Olofsson, Roman Grabic, Jerker Fick
ABSTRACT
The concentrations and behaviour of 105 different active pharmaceutical ingredients (APIs) in the aqueous phase of sewage water within a municipal sewer collection system have been investigated. Sewage water samples were gathered from seven pump stations (one of which was located within a university hospital) and from sewage water treatment influent and effluent. The targeted APIs were quantified using a multi-residue method based on online solid phase extraction liquid chromatography tandem mass spectrometry. The method was thoroughly validated and complies with EU regulations on sample handling, limits of quantification, quality control and selectivity. 51 APIs, including antibiotics, antidepressants, hypertension drugs, analgesics, NSAIDs and psycholeptics, were found frequently within the sewer collection system. API concentrations and mass flows were evaluated in terms of their frequency of detection, daily variation, median/minimum/maximum/average concentrations, demographic dissimilarities, removal efficiencies, and mass flow profiles relative to municipal sales data. Our results suggest that some APIs are removed from, or introduced to, the aqueous phase of sewage waters within the studied municipal collection system.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%