  • Search Results
  • An optically and thermally switchable electronic structure based on an anthracene-BODIPY conjugate.
An optically and thermally switchable electronic structure based on an anthracene-BODIPY conjugate.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-02-25)
Tatsuya Aotake, Mitsuharu Suzuki, Kazuaki Tahara, Daiki Kuzuhara, Naoki Aratani, Naoto Tamai, Hiroko Yamada
ABSTRACT
An optically and thermally responsive boron dipyrromethene (BODIPY) dye, namely, meso-2-(9,10-dihydro-9,10-ethanoanthracene-11,12-dione) (DK)-linked, bicyclo[2.2.2]octadiene (BCOD)-fused BODIPY (BCOD-DK), was synthesized. The weakly luminous structure of BCOD-DK can be changed quantitatively to that of the strongly fluorescent BODIPY BCOD-Ant by optical excitation at the DK unit, which induces double decarbonylation of the DK unit to give an anthracene unit. The solvent effect on the fluorescence properties of BCOD-DK suggests that the dramatic change in fluorescence intensity is controlled by intramolecular electron transfer from the BODIPY moiety to the meso-DK substituent. BCOD-DK is converted to meso-DK benzene-fused BODIPY (Benzo-DK) by heating at 220 °C with 64-70 nm redshift of absorption and fluorescence peaks without changing the fluorescence quantum yield of ΦF =0.08 in dichloromethane. Benzo-DK can be converted to strongly fluorescent meso-anthracene benzene-fused BODIPY Benzo-Ant by optical excitation. Thus, BCOD-DK can show four different optical performances simply by irradiation and heating, and hence may be applicable for optical data storage and security data encryption.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm