  • Search Results
  • Molecular variants of human papilloma virus 16 E2, E4, E5, E6 and E7 genes associated with cervical neoplasia in Romanian patients.
Molecular variants of human papilloma virus 16 E2, E4, E5, E6 and E7 genes associated with cervical neoplasia in Romanian patients.
Archives of virology (2014-08-22)
Adriana Plesa, Gabriela Anton, Iulia V Iancu, Carmen C Diaconu, Irina Huica, Anca D Stanescu, Demetra Socolov, Elena Nistor, Elena Popa, Mihai Stoian, Anca Botezatu
ABSTRACT
The aim of this study was to identify and associate the sequence variations of human Papillomavirus 16 (HPV16) genes from women who live in two different areas of Romania and associate them with malignant progression. One hundred twenty-four HPV16-positive cervical isolates were collected, and the E2, E4, E5, E6 and E7 viral genes were sequenced. Two new missense mutations in the E6 gene (C279G and A305C) were found (together or alone, in association with other mutations) in 44 of 124 cases. The most frequently simultaneously mutated genes were E4/E2 hinge, E5 and E6 (p = 0.0004) in squamous cell carcinoma (SCC) samples. Also, for SCC patients, the best-correlated mutation patterns were obtained for E4/E2 hinge-E5 (r = 0.7984; p < 0.0001). No sample was found to have all of the investigated viral genes concurrently mutated. Phylogenetic analysis was performed to characterize the viral variants. Similar results were found for SCC and cervical intraepithelial neoplasia III (CINIII) cases. After all of the target gene sequences were assembled, all patients were found to be infected with viruses of the HPV16- European-German (EG) lineage, and two clusters were identified, the first (55/96 variants) from Moldavia and the second (41/96 variants) from Bucharest. The distinct cluster derived from EG in Moldavia could partially explain the increased frequency of SCC in this area. This study has generated a comprehensive set of sequence variation data on HPV16 circulating in Romania to join the existing data and highlight the important role of HPV16 variants during cervical carcinogenesis.
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Supelco
Benalaxyl-M, PESTANAL®, analytical standard
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Boric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%