  • Search Results
  • Digital plethysmography and arginine metabolism in prehypertension: effect of nebivolol therapy.
Digital plethysmography and arginine metabolism in prehypertension: effect of nebivolol therapy.
Journal of clinical hypertension (Greenwich, Conn.) (2014-12-17)
Ramprasad Kandavar, Camilo Fernandez, Gary E Sander, Catherine Kim, Santhosh Velaga, Sergiy Sukhanov, Patrice Delafontaine, Peter Egan, Louise E Roffidal, Thomas D Giles
ABSTRACT
Prehypertension is an important phenotype for cardiovascular risk and development of established hypertension. To better understand the early circulatory changes in this group, the authors studied 34 patients with prehypertension (blood pressure 120-139/80-89 mm Hg) using digital plethysmography for measurement of blood flow and reactive hyperemic index (RHI). Arterial augmentation index (AI) was also measured. Because prehypertension is associated with endothelial dysfunction and decreased availability of nitric oxide (NO), indices of arginine metabolism (l-arginine, asymmetric dimethylarginine (ADMA), symmetric dimethylarginine, and l-citrulline) were measured. Nebivolol (5 mg/d), a vasodilating β1 -antagonist with β3 -agonist activity, was studied in a double-blind fashion for 8 weeks. Nebivolol increases the bioavailability of NO. Prehypertension was associated with normal RHI and baseline digital blood flow. AI was abnormal and associated with diastolic blood pressure. ADMA concentration was increased at baseline. After 8 weeks of nebivolol therapy, RHI, ADMA, symmetric dimethylarginine, and AI showed no significant change, but digital blood flow and l-citrulline levels were significantly increased. Prehypertension is associated with increased ADMA and evidence of increased arterial stiffness and preserved RHI. Nebivolol therapy is associated with digital vasodilation and increased NO production, as depicted by increased levels of l-citruline and mean digital blood flow.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free