  • Search Results
  • Quantification of trabectedin in human plasma: validation of a high-performance liquid chromatography-mass spectrometry method and its application in a clinical pharmacokinetic study.
Quantification of trabectedin in human plasma: validation of a high-performance liquid chromatography-mass spectrometry method and its application in a clinical pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2014-03-26)
Monique Zangarini, Laura Ceriani, Federica Sala, Elena Marangon, Renzo Bagnati, Maurizio D'Incalci, Federica Grosso, Massimo Zucchetti
ABSTRACT
A rapid, sensitive and specific HPLC-MS/MS method was developed and validated for the quantification of trabectedin in human plasma after using deuterated trabectedin as Internal Standard (IS). After the addition of ammonium sulphate, the analyte was extracted from human plasma with acidified methanol (0.1 M HCl). Chromatographic separation was done on an Accucore XL C₁₈ column (4 μm; 50 mm × 2.1 mm) using a Mobile Phase (MP) consisting of CH₃COONH₄ 10 mM, pH 6.8 (MP A) and CH₃OH (MP B). The mass spectrometer worked with electrospray ionization in positive ion mode and Selected Reaction Monitoring (SRM), using target ions at [M-H₂O+H]⁺ m/z 744.4 for trabectedin and [M-H₂O+H]⁺m/z 747.5 for the IS. The standard curve was linear (R² ≥ 0.9955) over the concentration range 0.025-1.0 ng/ml and had good back-calculated accuracy and precision. The intra- and inter-day precision and accuracy determined on three quality control samples (0.04, 0.08 and 0.80 ng/ml) were <9.9% and between 98.3% and 105.3%, respectively. The extraction recovery was >81% and the lower limit of quantification 0.025 ng/ml. The method was successfully applied to study trabectedin pharmacokinetics in a patient with a liposarcoma who received 1.3 mg/m² as a 24 h continuous i.v. infusion.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Ammonium acetate, suitable for mass spectrometry (MS), LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)