  • Search Results
  • Prevalence and genotyping of high risk human papillomavirus in cervical cancer samples from Punjab, Pakistan.
Prevalence and genotyping of high risk human papillomavirus in cervical cancer samples from Punjab, Pakistan.
Viruses (2014-07-19)
Abida Siddiqa, Maidah Zainab, Ishtiaq Qadri, Muhammad Faraz Bhatti, Joanna L Parish
ABSTRACT
Cervical cancer is the third most common cause of cancer-related death in women worldwide. Infection with high-risk human papillomavirus (HPV) is established as the cause of cervical carcinoma, therefore, high risk HPV detection may have prognostic significance for the women who are at increased risk of disease progression. The paucity of data on the incidence of cervical cancer in Pakistan makes it difficult to determine disease burden. Even less information is available regarding the prevalent HPV strains in cervical specimens collected from this region. Cervical cancer is a neglected disease in Pakistan in terms of screening, prevention, and vaccination. Identification and accurate genotyping of the virus burden in cancer specimens is important to inform intervention policies for future management of HPV associated disease and to potentially stratify patients dependent on HPV status. In this study, detection and genotyping of HPV types 16 and 18 from 77 cervical specimens were carried out. Consensus primers GP5+/GP6+, which detect 44 genital HPV types, and type specific primers (TS16 and TS18) were used in conjunction with newly designed type specific primers. Using a combination of these methods of detection, a total of 94.81% (95% CI ±4.95) of cervical lesions were positive for HPV. Single infections of HPV16 were detected in 24.68% (95% CI ±9.63) of total samples and HPV18 was found in 25.97% (95% CI ±9.79) samples. Interestingly, a high proportion of samples (40.26%, 95% CI ±10.95) was positive for both HPV16 and 18, indicating a higher incidence of co-infection than previously reported for similar ethnic regions. The HPV genotype of 3.90% of HPV positive samples remained undetected, although these samples were positive with the GP5+/GP6+ primer set indicating infection with an HPV type other than 16 or 18. These data indicate that the overall incidence of high risk HPV infection in cervical cancer and intraepithelial neoplasia specimens in Punjab, Pakistan is in line with the worldwide prevalence, but that the incidence of HPV16 and 18 co-infections in our cohort is higher than that previously reported.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis