  • Search Results
  • Pre-rigor temperature control of Chinese yellow cattle carcasses to 12-18 °C during chilling improves beef tenderness.
Pre-rigor temperature control of Chinese yellow cattle carcasses to 12-18 °C during chilling improves beef tenderness.
Meat science (2014-12-03)
Yuqing Liu, Yanwei Mao, Yimin Zhang, Rongrong Liang, Renhuan Wang, Lixian Zhu, Xianyong Meng, Xin Luo
ABSTRACT
This study evaluates the effects of pre-rigor temperature control on quality traits of Chinese yellow cattle M. longissimus lumborum (LL). One stepwise chilling (SC) treatment was used on one half-carcass, involved a fast chilling (-11 ± 1 °C;0.5 m/s) for 2h, then the refrigeration was stopped to hold a core temperature of 12-18 °C until 10h postmortem, followed by a 1 ± 1 °C chilling (0.5 m/s) to 48h postmortem. The other half-carcass was conventional chilled at 1 ± 1 °C (0.5 m/s) until 48h as control chilling (CC). Quality attributes were evaluated at 1, 7 and 14 days. The SC treatment resulted in decreased WBSF and increased myofibril fragmentation index compared with control. SC-treated LL at 7d postmortem had a lower WBSF than those of CC-treated at 14d. This pre-rigor temperature controlled chilling is a realistic alternative for the beef industry in China to ensure adequate tenderness and shorten aging time.
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, Vetec, reagent grade, 99%
Supelco
Sucrose, analytical standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard