  • Search Results
  • Factors affecting the adsorptive removal of bisphenol A in landfill leachate by high silica Y-type zeolite.
Factors affecting the adsorptive removal of bisphenol A in landfill leachate by high silica Y-type zeolite.
Environmental science and pollution research international (2014-09-12)
Xiaoqiang Chen, Taku Fujiwara, Shuji Fukahori, Tomonori Ishigaki
ABSTRACT
Although bisphenol A (BPA), a representative endocrine-disrupting compound, has been detected frequently in landfill leachate, effective technologies for BPA removal from landfill leachates are limited. We used high silica Y-type zeolite (HSZ-385) for the selective adsorption of BPA from landfill leachate, and factors affecting this adsorption are discussed. Higher removal efficiencies at pH 5.0-9.0 imply that neutral BPA is adsorbed more easily onto HSZ-385 than monomeric or divalent BPA anions. An increase in ionic strength and sodium acetate concentration did not affect BPA adsorption significantly, while the removal efficiency decreased slightly when more than 50 mgC/L of humic acid was added. HSZ-385 was applied to synthetic leachates that simulate the composition of landfill leachate at various degradation stages. In young acidic leachates that contain sodium acetate, the use of HSZ-385 for the adsorptive removal of BPA appears to be more effective than in old alkaline leachates, which contain large amounts of humic acid. In addition, 82 % BPA removal was achieved from young raw leachates using HSZ-385, which demonstrates that selective BPA removal from actual landfill leachate has been achieved.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%