  • Search Results
  • C-type lectin-like domain and fibronectin-like type II domain of phospholipase A(2) receptor 1 modulate binding and migratory responses to collagen.
C-type lectin-like domain and fibronectin-like type II domain of phospholipase A(2) receptor 1 modulate binding and migratory responses to collagen.
FEBS letters (2015-03-01)
Soichiro Takahashi, Kazuhiro Watanabe, Yosuke Watanabe, Daisuke Fujioka, Takamitsu Nakamura, Kazuto Nakamura, Jun-ei Obata, Kiyotaka Kugiyama
ABSTRACT
Phospholipase A2 receptor 1 (PLA2R) mediates collagen-dependent migration. The mechanisms by which PLA2R interacts with collagen remain unclear. We produced HEK293 cells expressing full-length wild-type PLA2R or a truncated PLA2R that lacks fibronectin-like type II (FNII) domains or several regions of C-type lectin-like domain (CTLD). We show that the CTLD1-2 as well as the FNII domain of PLA2R are responsible for binding to collagen and for collagen-dependent migration. Thus, multiple regions and domains of the extracellular portion of PLA2R participate in the responses to collagen. These data suggest a potentially new mechanism for PLA2R-mediated biological response beyond that of a receptor for secretory PLA2.
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)