  • Search Results
  • Fast and neat--determination of biochemical quality parameters in cocoa using near infrared spectroscopy.
Fast and neat--determination of biochemical quality parameters in cocoa using near infrared spectroscopy.
Food chemistry (2015-03-22)
Andrea Krähmer, Annika Engel, Daniel Kadow, Naailah Ali, Pathmanathan Umaharan, Lothar W Kroh, Hartwig Schulz
ABSTRACT
The qualitative heterogeneity and increasing consumption of cocoa products require fast and efficient methods for quality assessment of fermented cocoa with regard to fermentation quality and flavor potential. To date, quality control is achieved by visual inspection (e.g., "cut test") and sensory testing. Chromatographic methods for quantification of flavor relevant substances are limited in their applicability in standard quality control due to laborious isolation and purification steps. Therefore, the aim of this study was the development of a near infrared spectroscopic (NIRS) method for routine analytical prediction of biochemical quality parameters. Different compound classes like phenolic substances (R(2)=0.93) or organic acids (R(2)=0.88) as well as individual substances like epicatechin (R(2)=0.93) or lactic acid (R(2)=0.87) could be precisely determined just as fermentation time (R(2)=0.92) and pH value (R(2)=0.94) presenting NIRS as fast and reliable alternative in routine quality assessment.
Sigma-Aldrich
D-(−)-Fructose, BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
D-(−)-Fructose, BioXtra
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
D-(−)-Fructose, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99%
USP
Fructose, United States Pharmacopeia (USP) Reference Standard
Fructose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, Vetec, reagent grade, 99%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard