  • Search Results
  • Synthesis of a molecularly imprinted polymer to isolate glucosamine from plant extracts by an ionic-non-covalent dual approach.
Synthesis of a molecularly imprinted polymer to isolate glucosamine from plant extracts by an ionic-non-covalent dual approach.
International journal of cosmetic science (2014-11-18)
N Henry, P Favetta, R Delépée, J-M Seigneuret, L A Agrofoglio
ABSTRACT
The objective of this study was to synthesize a novel glucosamine-imprinted sorbent based on ionic and non-covalent dual approach to purify glucosamine from chicory root extracts. The synthesis of the molecularly imprinted polymer was optimized in terms of choice of monomers, porogen, cross-linker and initiator to have the best recognition as possible for targeted molecule. The sorbent obtained was characterized by nitrogen sorption (BET), scanning electron microscopy (SEM) and solid-phase extraction (SPE) to plot adsorption isotherms. The selectivity of polymer between glucosamine and interfering salt as ammonium sulphate was calculated. Extraction procedure was optimized in terms of loading, washing and elution solvents, to have the best recovery of glucosamine. Compounds were analysed by HPLC-UV after chemical derivatization. The results showed that the optimal conditions of extracting glucosamine on this new type of sorbent were as follows: percolation of plant extract in EtOH/aqueous HCl pH 3, washing of cartridge with water and elution of compound of interest with aqueous acetic acid solution at 5%. The recoveries of glucosamine were around 53% and 70%, from aqueous standard solution and aqueous chicory roots extracts, respectively, on the molecularly imprinted polymer. And, only 11% and 7% of the ammonium sulphate were recovered from standard solution and chicory roots extract, respectively. The use of the MIP as solid-phase extraction sorbent was able to extract preferentially glucosamine from structural analogues and ammonium salt. Assays on chicory roots extracts were carried out, and the MIP showed good results allowing the transfer methodology at semi-industrial scale for cosmetic companies. The optimized protocol of extraction of glucosamine allowed using only eco-friendly solvents, as ethanol, water and acetic acid.
Product Description
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Supelco
Methyl acetoacetate, analytical standard
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Sodium persulfate, BioUltra, ≥99%
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, 99%
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Sodium persulfate, BioXtra, ≥99%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)
Glucosamine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium sulfate, Vetec, reagent grade, 99%
USP
Glucosamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium persulfate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Supelco
Glucosamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Glucosamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium persulfate, purum p.a., ≥98% (RT)
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid, 99%